Learn More
Medchemexpress LLC D-sorbitol-d8 | 287962-59-8 | MFCD02683970 | 98.0% | 190.22 | C6H6D8O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0400S10MG
Encompass_Preferred
D-Sorbitol-d8 is the deuterium-labeled form of D-sorbitol supplied as a small-mass research standard for use in analytical and metabolic studies. It is intended for use as a stable-isotope internal standard, tracer, or labeled reagent in techniques such as mass spectrometry and metabolite profiling.
  • Deuterium-labeled (d8) D-sorbitol for isotope-labeled analyses.
  • Suitable as a stable-isotope internal standard for mass spectrometry.
  • Provided as a powder in small milligram pack sizes for analytical workflows.
  • Chemical formula C6H6D8O6; molecular weight 190.22.
Catalog No. 50-002-98929