Learn More
Medchemexpress LLC D-(+)-Talose-13C-1 | 83379-36-6 | 99.0% | 181.15 g·mol⁻1 | C6H12O6 | 25 MG

Supplier: Medchemexpress LLC HYW145492S325MG
D-(+)-Talose-13C-1 is the carbon-1 13C-labeled isotopologue of D-talose, supplied as a high-purity solid for use as a tracer and as an internal standard in analytical methods such as NMR, GC-MS, and LC-MS.
- Labeled at carbon-1 with 13C for tracer studies.
- Suitable as an internal standard for NMR, GC-MS, and LC-MS.
- High purity: 99.0%.
- Pack size appropriate for analytical applications.
- Molecular formula C6H12O6; molecular weight 181.15 g·mol⁻1.
- CAS number 83379-36-6 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.