Learn More
Medchemexpress LLC D-(+)-talose-13C-1 | 83379-36-6 | MFCD08459988 | 99.0% | 181.15 g/mol | C5[13C]H12O6 | 100 MG

Supplier: Medchemexpress LLC HYW145492S3100MG
D-(+)-Talose-13C-1 is the 13C-labeled form of D-(+)-talose provided for research use as a white to off-white solid. It has a molecular weight of 181.15 g/mol and a reported purity of 99.0%. This material is intended for use as a tracer or internal standard in analytical and biochemical applications; consult storage and stability guidelines before use.
- Contains a single 13C isotopic label for tracer studies.
- High purity suitable for analytical standards and internal standard use.
- Solid, white to off-white appearance for easy handling.
- Molecular weight 181.15 g/mol; molecular formula C5[13C]H12O6.
- Powder storage: stable at -20°C for extended periods; in solvent stability is limited.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.