Learn More
Medchemexpress LLC D-Threitol | 2418-52-2 | 99.95% | 122.12 | 25 MG

Supplier: Medchemexpress LLC 25MGHYW012846
D-threitol serves as an antifreeze agent in the Alaskan beetle *Upis ceramboides*. It is classified as an endogenous metabolite and a saccharide, appearing as a white to light yellow solid. This product is for research use only and not sold to patients.
- Targets human endogenous metabolite
- Available as a powder
- Can be stored at -20°C for 3 years in powder form
- Soluble in DMSO at 250 mg/mL
- Multiple in vivo dissolution methods available for clear solutions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.