Learn More
Medchemexpress LLC D-Threitol | 2418-52-2 | 99.95% | 122.12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0128465MG
Encompass_Preferred
D-threitol serves as an antifreeze agent in the Alaskan beetle Upis ceramboides. It is an endogenous metabolite and a saccharide.
  • Endogenous metabolite
  • Saccharide
  • Serves as an antifreeze agent
Catalog No. 50-004-54846