Learn More
Medchemexpress LLC D-threo-PDMP 10mM 1mL | 109836-82-0 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY116392E10MM 1ML
Encompass_Preferred
D-threo-PDMP is a potent glucosylceramide synthase inhibitor supplied as a ready-to-use 10 mM solution in DMSO for use in biochemical and cell-based research investigating glycosphingolipid metabolism, neurite growth, and apoptosis pathways.
  • Ready-to-use 10 mM solution in DMSO for immediate application
  • High chemical purity suitable for research assays
  • Effective inhibitor of glucosylceramide synthase activity
  • Stable when stored cold and protected from moisture
  • Commonly used in studies of glycosphingolipid-related cellular processes
Catalog No. 50-002-28407