Learn More
Medchemexpress LLC D-threo-PDMP 50mg | 109836-82-0 | 390.56 g/mol | C23H38N2O3 | 50 MG

Supplier: Medchemexpress LLC HY116392E50MG
D-threo-PDMP is a research compound that inhibits glucosylceramide synthase (GCS) and is used to modulate glycosphingolipid metabolism in biochemical and pharmacological studies. The compound is supplied as a colorless to off-white oil with high reported purity and is offered in small research pack sizes.
- Potent glucosylceramide synthase inhibitor for research use.
- High purity (99.50%) for reliable experimental results.
- Available in small quantities, including 50 MG research packs.
- Soluble in DMSO and available as a 10 mM solution option.
- Recommended storage at -20°C under nitrogen to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.