Learn More
Medchemexpress LLC D-threo-PDMP hydroch 10mg | 139889-62-6 | 427.02 | C23H39ClN2O3 | 10 MG

Supplier: Medchemexpress LLC HY116392F10MG
D-threo-PDMP hydrochloride is the hydrochloride salt of D-threo-PDMP, a glucosylceramide synthase inhibitor used as a research reagent to study sphingolipid metabolism and glycosphingolipid-related cellular effects.
- Inhibits glucosylceramide synthase, reducing glycosphingolipid synthesis.
- Purity ≥98.0%.
- Molecular formula C23H39ClN2O3; molecular weight 427.02 g·mol-1.
- White to off-white solid appearance suitable for laboratory handling.
- Available in small research pack sizes, including 10 mg.
- Recommended storage: sealed, away from moisture; store solid at -20°C; in solvent: -80°C up to 6 months, -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.