Learn More
Cayman Chemical D-threo-PPMPhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2267710 MG
Encompass_Preferred
A GlyCer synthetase inhibitor; an inhibitor of MDCK cell growth in vitro (70% reduction at 20 µM); an inhibitor of DNA synthesis at 3 µM
Catalog No. 50-296-4551