Learn More
Cayman Chemical d-TNFR2 M196R varInthumn 100ug
SDP
Supplier:  Cayman Chemical 32023100 UG
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged DS-TNFR2 M196R variant expressed in HEK293 cells • Amino acids: 23-268 • MW: 27.7 kDa
Catalog No. 50-294-8434