Learn More
Medchemexpress LLC D-(+)-Trehalose | 99-20-7 | 342.30 | 250 MG

Supplier: Medchemexpress LLC 250MGHYN1132R
D-(+)-Trehalose (α,α-Trehalose) is an orally active disaccharide that exhibits anti-desiccant and cryopreservative activities. It functions as an osmolyte and stress protectant, aiding in the storage and transport of carbon. D-(+)-Trehalose is suitable for use as a food ingredient and pharmaceutical excipient.
- Orally active disaccharide
- Anti-desiccant activity
- Cryopreservative activity
- Acts as an osmolyte and stress protectant
- Helps in the storage and transport of carbon
- Can be used as a food ingredient
- Can be used as a pharmaceutical excipient
- Purity 99.88% (HPLC)
- Appearance: solid
- Color: white to off-white
- No toxic effects on Chinese hamster ovary (CHO) cells at 5000 μg/mL for 21 hours
- Mild, self-limiting side effects in mice and rats at high doses (1-16 g/kg), without systemic toxicity
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.