Learn More
Alkali Scientific D (+) Trehalose Dihydrate, 100 G
SDP
Supplier:  Alkali Scientific DTR001CP
Encompass_Preferred
D (+) Trehalose Dihydrate is a stabilizing carbohydrate compound used in various laboratory applications, particularly for cell protection and preservation. Trehalose is a naturally occurring disaccharide sugar known for its ability to stabilize proteins and cellular structures during stress conditions. The compound helps protect cells from damage caused by dehydration, freezing, or oxidative stress. This product is commonly used in the preservation of biological samples, enzyme activity studies, and pharmaceutical applications. Available in a 100g bottle, it is ideal for research and biochemical processes that require stabilization of cellular components or microbial cultures.
Catalog No. 50-006-60173