Learn More
Medchemexpress LLC D-Trehalose dihydrate | 6138-23-4 | 378.33 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN1132AR
Encompass_Preferred
D-(+)-Trehalose dihydrate (Standard) is the analytical standard of D-(+)-Trehalose dihydrate. This product is intended for research and analytical applications. Isolated from *Saccharomyces cerevisiae*, it can be used as a food ingredient and pharmaceutical excipient.
  • Intended for research and analytical applications
  • Serves as an analytical standard and reference standard for assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-89852