Learn More
Bachem (D-Trp6)-LHRH Acetate
SDP
Supplier:  Bachem 4033016.0005
Encompass_Preferred
(D-Trp6)-LHRH Acetate, Pyr-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-Gly-NH2, 57773-63-4, C64H82N18O13, Mr 1311.47, 5mg. (D-Trp6)-GnRH, Triptorelin acetate salt, [DTrp6] LH-RH, amide. LHRH agonist used in the treatment of prostate cancer. Triptorelin decreases the secretion of LH and FSH. CAS-number (triptorelin acetate) 140194-24-7.
Catalog No. 50-259-804