Learn More
Medchemexpress LLC D-Tryptophan, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 86123-11-7 | 100.0% | 426.46 | 25 G

Supplier: Medchemexpress LLC 25GHYW008963
Fmoc-D-Trp-OH (Nα-Fmoc-D-tryptophan) is a biochemical reagent suitable for life science research. This organic compound has a molecular weight of 426.46 and appears as a white to off-white solid with high purity. It is soluble in DMSO.
- Biochemical reagent for life science research.
- Organic compound with a molecular weight of 426.46.
- Appears as a white to off-white solid.
- Purity of 99.98% (ee.: 100.00%).
- Soluble in DMSO at concentrations of ≥ 100 mg/mL.
- Stable in powder form at -20°C for 3 years or 4°C for 2 years.
- Stable in solvent at -80°C for 6 months or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.