Learn More
Medchemexpress LLC D-tyrosine, N-[(9H-fluoren-9-yl)methoxycarbonyl]- | 112883-29-1 | 99.5% | 403.43 g/mol | C24H21NO5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0487035G
Encompass_Preferred
(((9H-Fluoren-9-yl)methoxy)carbonyl)-D-tyrosine (Fmoc-D-tyrosine) is an Fmoc-protected D-tyrosine used as a protected amino acid building block for solid-phase peptide synthesis and related research applications. It is a white to off-white solid with molecular formula C24H21NO5, molecular weight 403.43 g/mol, and typical purity 99.5%.
  • Fmoc-protected N-terminal protection for solid-phase peptide synthesis.
  • D-configuration for incorporation of D-amino acids in peptides.
  • High purity suitable for peptide synthesis and analytical work.
  • White to off-white solid form, convenient for handling and storage.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-09856