Learn More
Medchemexpress LLC D-val-leu-arg-pNA acetate | 162303-66-4 | 99.8% | 566.65 g/mol | C25H42N8O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY114150D5MG
Encompass_Preferred
D-Val-Leu-Arg-pNA acetate is a synthetic peptide substrate for kallikrein proteases that releases p-nitroaniline upon enzymatic cleavage, enabling colorimetric measurement of protease activity. Supplied as a white to off-white solid with high purity, it is intended for biochemical assays and enzyme kinetics studies with recommended frozen storage for stability.
  • Used as a substrate for kallikrein activity assays
  • Colorimetric detection via p-nitroaniline release
  • High purity (≈99.8%) for reliable assay performance
  • Suitable for enzyme kinetics and activity screening
  • Stable when stored frozen; follow recommended storage conditions
Catalog No. 50-002-63881