Learn More
Cayman Chemical D-Val-Leu-Lys-pNA 1mg
SDP
Supplier:  Cayman Chemical 365801 MG
Encompass_Preferred
A colorimetric plasmin substrate; hydrolysis releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of plasmin activity
Catalog No. 50-291-3910