Learn More
Medchemexpress LLC D-Valine | 640-68-6 | 117.15 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN0717A5G
Encompass_Preferred
D-Valine is the enantiomer of L-Valine, one of 20 proteinogenic and essential amino acids. It is intended for research use only.
  • Can be used to prevent the growth of fibroblasts in primary and secondary cell cultures of bovine artery endothelial cells
  • Inhibits biofilm formation of P. gingivalis
Catalog No. 50-004-16701