Learn More
Medchemexpress LLC D-valine-d8 | 1116448-82-8 | 99.1% | 125.20 g/mol | C5H3D8NO2 | 10 MG

Supplier: Medchemexpress LLC HYN0717AS10MG
D-Valine-d8 is the deuterium-labeled enantiomer of valine used as an isotopically labeled standard or tracer in analytical and biochemical applications, including mass spectrometry, metabolic tracing, and chiral studies. It is supplied as a solid with high isotopic purity for reliable quantification.
- Deuterium-labeled D-valine suitable for isotopic labeling and internal standards.
- Molecular formula C5H3D8NO2 and molecular weight 125.20 g/mol.
- High purity (99.1%) for accurate analytical results.
- Available as a solid in small mg packages, including 10 MG.
- CAS number 1116448-82-8 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.