Learn More
Medchemexpress LLC D-valine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-3-[(triphenylmethyl)thio]- | 201532-01-6 | >95.0% | 613.76 g/mol | C39H35NO4S | 5 G

Supplier: Medchemexpress LLC HYW0486975G
Fmoc-D-Pen(Trt)-OH is an Fmoc-protected D-penicillamine derivative used in peptide synthesis, notably for preparing cyclic lanthionine enkephalin analogs.
- Fmoc-protected D-penicillamine with trityl side-chain protection.
- Molecular formula C39H35NO4S.
- Molecular weight 613.76 g/mol.
- Purity ≥95.0% (NMR).
- CAS number 201532-01-6.
- Used in solid-phase peptide synthesis and modified peptide preparation.
- Available in 1 g, 5 g, 10 g, and 50 g pack sizes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.