Learn More
Medchemexpress LLC α-D-Xylose | 6763-34-4 | 150.13 | 500 G
SDP
Supplier:  Medchemexpress LLC HYN2018500G
Encompass_Preferred
α-D-Xylose, also known as α-D-Xylopyranose, is an endogenous metabolite and a basic component of the five-carbon fraction of biomass. It acts as a precursor of hemicellulose and participates in various enzyme-catalyzed reactions within metabolic pathways. This compound is also utilized in industries such as tanning and dyeing, and it finds application as a diabetic food.
  • Basic component of biomass five-carbon fraction
  • Precursor of hemicellulose
  • Participates in enzyme-catalyzed reactions and metabolic pathways
  • Used in tanning, dyeing, and as a diabetic food
  • Appears as a solid, white to off-white substance
  • Functions as an endogenous metabolite
Catalog No. 50-004-15996