Learn More
Medchemexpress LLC D-Xylulose-2-13C | 131771-47-6 | 99.5% | 151.12 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW010256S1
Encompass_Preferred
D-Xylulose-2-13C is the 13C labeled D-Xylulose. D-xylulose serves as a precursor for the pentiol D-arabi. This product is intended for research use only and is not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration has garnered attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Is a 13C labeled D-Xylulose.
  • Can be utilized as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66710