Learn More
Medchemexpress LLC D13-9001 | 957471-96-4 | 99.9% | 693.78 | C31H39N11O6S | 5MG
SDP
Supplier:  Medchemexpress LLC HY1248195MG
Encompass_Preferred
D13-9001 is a small-molecule inhibitor of bacterial efflux pump subunits AcrB and MexB, supplied for research applications to investigate efflux-mediated antibiotic resistance. The compound has a molecular formula of C31H39N11O6S, a molecular weight of 693.78, and is offered at high reported purity for in vitro studies.
  • Inhibits AcrB and MexB efflux pump subunits.
  • Supports studies of antibiotic resistance mechanisms.
  • High purity suitable for analytical and biological assays.
  • Molecular weight 693.78 and formula C31H39N11O6S.
  • Available in small-molecule pack sizes including 5 mg.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-38875