Learn More
Biolegend D2-40 Lymphatic Endothelial Ma
Supplier:  Biolegend 916602
Encompass_Preferred
D2-40 Lymphatic Endothelial Marker mAb [D2-40]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: IHC; Size: 1 mL
Catalog No. 50-272-1624