Learn More
Cole-Parmer D2 DET LAMP TSP UV100 150 200
Supplier:  Cole-Parmer 1194129
Encompass_Preferred
Cole-Parmer Deuterium (D2) Detector Lamp for TSP UV100, 150, 200, 1000, 2000, 3000.standard life 1000 hour bulb
Catalog No. NC2682086