Learn More
Cayman Chemical D2PMhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1116010 MG
Encompass_Preferred
A psychoactive designer drug; also used in organic synthesis to prepare the Corey-Bakshi-Shibata catalyst; intended for forensic purposes
Catalog No. 50-295-5796