Learn More
Selleck Chemical LLC D5 epichlorohydrin E8222-5MG
SDP
Supplier:  Selleck Chemical LLC E82225MG
Encompass_Preferred
(D5)epichlorohydrin is the deuterated form of epichlorohydrin Epichlorohydrin is a highly toxic industrial raw material with alkylating activity
Catalog No. 50-000-41528