Learn More
SPECTRUM QUALITY STANDARDS 5769INT-T-5 [w/4 Internals] (5×10 ml)
Supplier:  SPECTRUM QUALITY STANDARDS 5769INTT5
Encompass_Preferred
Internal standards containing Deuterated Ethylbenzene-d10, Naphthalene-d8, Benzene-d6, and Toluene-d8 as specified in the ASTM method.
Catalog No. NC0996863