Learn More
Grainger D6075 Inventory Control Label Pink PK1000
Supplier:  Grainger 16U936
Encompass_Preferred
The Grainger 16U936 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning applications. This 8" x 4" x 1/2" hand pad has an open web construction that allows it to conform to curved surfaces. It is made of high-quality fibers that are durable and long-lasting. The pad is designed for light-duty scrubbing and cleaning of surfaces. It can be used wet or dry on a variety of materials including metal, plastic, glass and ceramic. The open web construction allows dirt and grime to pass through the pad during cleaning.
Catalog No. 50-276-7602