Learn More
Medchemexpress LLC HY-15162A 1mg Medchemexpress, D8-MMAE CAS:2070009-72-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15162A1MG
Encompass_Preferred
Medchemexpress, HY-15162A 1mg D8-MMAE CAS:2070009-72-0 D8-MMAE (D8-Monomethyl auristatin E) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9088