Learn More
Biocare Medical, LLC Da Vinci Green Diluent - 25 ml
SDP
Supplier:  Biocare Medical, LLC PD900H
Encompass_Preferred
Da Vinci Green antibody diluent is a modified PBS formulation with protein carrier and preservative. This modified PBS is formulated for superior performance and stability. In addition, it is protease and IgG-free. Da Vinci Green is Biocare's new standard-universal PBS diluent. It can also be used on an automated staining system.
Catalog No. 50-829-73