Learn More
Biocare Medical, LLC DAB SUBSTRATE BUFFER 25ML
SDP
Supplier:  Biocare Medical, LLC DS854H
Encompass_Preferred
DAB is a permanent chromogen that produces a brown stain in the presence of horseradish peroxidase. This two-component system consists of a liquid stable DAB chromogen and DAB substrate buffer, DAB Chromogen sold seperately.
Catalog No. 50-828-25