Learn More
Medchemexpress LLC Dabco-bis(sulfur dioxide) | 119752-83-9 | MFCD22377802 | 97.0% | 240.31 g/mol | C6H12N2O4S2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW03304910G
Encompass_Preferred
DABCO-Bis(sulfur dioxide) (DABSO) is a bench-stable, crystalline adduct used as a convenient solid source of sulfur dioxide for organic synthesis. It enables safe, controlled introduction of SO2 into laboratory reactions, supporting the synthesis of sulfonyl-containing compounds such as sulfonamides and sulfamides. The material is intended for research use and is supplied with supporting documentation including a safety data sheet and certificate of analysis.
  • Bench-stable solid source of sulfur dioxide for synthesis.
  • Enables preparation of sulfonamides and sulfamides.
  • More convenient and safer to handle than gaseous SO2.
  • High purity suitable for research applications.
  • Available in multiple laboratory pack sizes for scalability.
Catalog No. 50-002-99463