Learn More
Medchemexpress LLC HY-D1046 25mg Medchemexpress, Dabcyl acid, SE CAS:146998-31-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD104625MG
Encompass_Preferred
Medchemexpress, HY-D1046 25mg Dabcyl acid, SE CAS:146998-31-4 Dabcyl acid, SE is the amino-reactive form of Dabcyl acid (DABCYL), and widely used to prepare a variety of FRET-based probes that contain DABCYL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2752