Learn More
Medchemexpress LLC Dabigatran-d3 | 1246817-44-6 | MFCD21363573 | 95.1% | 474.53 g/mol | C25H22D3N7O3 | 500 UG

Supplier: Medchemexpress LLC HY10163S1500UG
Dabigatran-d3 is the deuterated analog of the anticoagulant dabigatran, provided for use as an internal standard in bioanalytical and pharmaceutical testing. It is intended to improve accuracy and precision in mass spectrometry quantitation workflows.
- Deuterated internal standard for LC-MS/MS quantification.
- High isotopic enrichment (100%).
- Chemical purity confirmed by HPLC (95.1%).
- Molecular formula C25H22D3N7O3 and molecular weight 474.53 g/mol.
- Suitable for pharmacokinetic, bioanalytical, and method validation studies.
- Supplied as an off-white solid with an accompanying certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.