Learn More
TARGETMOL CHEMICALS INC DABIGATRAN ETEXILATE MES 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5133100MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dabigatran Etexilate Mesylate (BIBR 1048MS) is the orally active prodrug of dabigatran. Dabigatran is a reversible and selective direct thrombin inhibitor (DTI Ki 4.5 nM). purity: 99%
Catalog No. 50-257-6709