Learn More
Selleck Chemical LLC Dabigatran etexilate mesylate
SDP
Supplier:  Selleck Chemical LLC S596025MG
Encompass_Preferred
Dabigatran etexilate mesylate (Pradaxa BIBR 1048MS Dabigatran etexilate methanesulfonate) is an orally active prodrug of Dabigatran which is a reversible and selective direct thrombin inhibitor (DTI) Dabigatran selectively and reversibly inhibits human thrombin with Ki of 4 5 nM and thrombin-induced platelet aggregation with IC50 of 10 nM
Catalog No. 50-000-03412