Learn More
Medchemexpress LLC Dabigatran ethyl ester hydrochloride | 211914-50-0 | 99.4% | C27H30ClN7O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY7752110MM/1ML
Encompass_Preferred
Dabigatran ethyl ester hydrochloride is a potent inhibitor of ribosyldihydronicotinamide dehydrogenase (NQO2) with an IC50 value of 0.8 μM. It also functions as a thrombin inhibitor. The ethyl ester group enhances interaction with hydrophobic amino acids, contributing to its higher affinity for both thrombin and NQO2 compared to Dabigatran. This compound is a selective, reversible, and potent thrombin inhibitor, orally available as the prodrug, dabigatran etexilate.
  • Potent inhibitor of ribosyldihydronicotinamide dehydrogenase (NQO2)
  • Thrombin inhibitor
  • Higher affinity for thrombin and NQO2 compared to Dabigatran
  • Selective and reversible thrombin inhibitor
  • Orally available as the prodrug dabigatran etexilate
  • For research use only
Catalog No. 50-004-33380