Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dabigatran impurity 75 | 1408238-36-7 | 479.57 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW00436610MG

Encompass_Preferred

Dabigatran impurity 75 is a laboratory chemical for research use only, intended for experienced personnel. This product is for the manufacture of substances and requires handling by suitably qualified scientists in equipped facilities. It is stable under recommended storage conditions but is harmful if swallowed, causes skin and serious eye irritation, and may cause respiratory irritation.

  • Appearance: Off-white to yellow solid
  • Purity: ≥97.0% (NMR)
  • Melting point: 119-135 °C
  • Recommended storage: 4°C, protect from light
  • In solvent storage: -80°C for 6 months, -20°C for 1 month, protect from light
  • Intended for research use only

Catalog No. 50-004-46954


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.