Learn More
Medchemexpress LLC Dabigatran impurity 75 | 1408238-36-7 | 479.57 | 5 MG

Supplier: Medchemexpress LLC HYW0043665MG
Dabigatran impurity 75 is an off-white to yellow solid, suitable for research applications. It has a high purity of ≥97.0% (NMR) and a molecular weight of 479.57. This compound should be stored at 4°C, protected from light, and in solvent at -80°C for 6 months or -20°C for 1 month, also protected from light.
- Chemical name: 1H-Benzimidazole-5-carboxylic acid, 2-[[[4-[[[(hexyloxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-, ethyl ester
- CAS number: 1408238-36-7
- Molecular formula: C26H33N5O4
- Molecular weight: 479.57
- Purity: ≥97.0% (NMR)
- Appearance: Off-white to yellow solid
- Storage conditions: 4°C, protected from light; in solvent: -80°C, 6 months; -20°C, 1 month (protected from light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.