Learn More
Medchemexpress LLC Dabigatran impurity 8 | 1408238-40-3 | MFCD27996077 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY2160910MG
Encompass_Preferred
Dabigatran impurity 8 is a characterized chemical impurity of the anticoagulant dabigatran intended for research and analytical use. It is supplied as a solid reference/intermediate for method development, impurity profiling, and synthetic studies. Users should refer to the certificate of analysis for storage and handling instructions; not for clinical or human use.
  • Chemical reference for impurity profiling and analytical method development.
  • Supplied as a stable powder suitable for storage under CoA-recommended conditions.
  • Characterized by CAS number 1408238-40-3, molecular formula C34H40N6O6, and molecular weight 628.72 g/mol.
  • Intended for research use only; not for clinical or human use.
  • Available in milligram pack sizes for analytical or synthetic applications.
Catalog No. 50-002-97541