Learn More
Medchemexpress LLC Dacinostat | 404951-53-7 | MFCD09970778 | 98.5% | 379.45 g/mol | C22H25N3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY136065MG
Encompass_Preferred
Dacinostat is a potent histone deacetylase (HDAC) inhibitor used in cancer and autophagy research. It exhibits low-nanomolar potency (HDAC IC50 ≈ 32 nM; HDAC1 IC50 ≈ 9 nM) and is supplied as a high-purity powder for laboratory studies.
  • High potency against HDAC enzymes, enabling low-nanomolar activity.
  • Supplied as a powder suitable for in vitro and cell-based studies.
  • High purity (98.5%), appropriate for biochemical assays.
  • Available in small research pack sizes suitable for screening and follow-up experiments.
  • Stable under recommended storage conditions for long-term archival.
  • Soluble in DMSO; use freshly opened solvent and aliquot stocks to avoid degradation.
  • Recommended handling for in vivo work: prepare fresh working solutions and limit DMSO to ≤2% when appropriate.
Catalog No. 50-002-22539