Learn More
TARGETMOL CHEMICALS INC Dacinostat 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T24545MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. LAQ824 (Dacinostat (NVP-LAQ824)) is a novel HDAC inhibitor with IC50 of 32 nM and is an activator of the p21 promoter. Purity 99.77%
Catalog No. 50-287-7623