Learn More
Selleck Chemical LLC Daclatasvir Dihydrochloride S5062-1g
SDP
Supplier:  Selleck Chemical LLC S50621G
Encompass_Preferred
Daclatasvir is an orally available antiviral agent that inhibits the NS5A region of the hepatitis C virus (HCV) used for the treatment of chronic HCV genotype 1 and 3 infection
Catalog No. 50-311-2458