Learn More
Medchemexpress LLC Daclatasvir (dihydrochloride) | 1009119-65-6 | MFCD25541736 | 99.8% | 811.80 | C40H52Cl2N8O6 | 50 MG

Supplier: Medchemexpress LLC HY10465R50MG
Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent hepatitis C virus (HCV) nonstructural protein 5A (NS5A) inhibitor. Supplied as a high-purity research reagent, it is intended for in vitro and preclinical studies only and not for human use.
- High purity: 99.8%.
- Molecular formula: C40H52Cl2N8O6.
- Molecular weight: 811.80 g/mol.
- Potent NS5A inhibitor with low-picomolar activity in biochemical assays.
- Supplied in laboratory-scale quantities suitable for reference standards and assay development.
- Documentation includes a product data sheet and safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.