Learn More
Cayman Chemical Daclatasvr 5mg
SDP
Supplier:  Cayman Chemical 237305 MG
Encompass_Preferred
An NS5A inhibitor (Kds = 8 and 210 nM for the NS5A33-202 and NS5A26-202 residues of HCV genotype 1b, respectively); potently inhibits HCV replication in multiple HCV replicon genotypes (EC50s = 9-146 pM) with the highest potency in genotypes 1b and 4a (EC50s = 9 and 12 pM, respectively); disrupts NS5A subcellular localization when used at a concentration of 1 nM; inhibits HCV RNA synthesis and virion assembly and secretion when used at a concentration of 1 nM; inhibits OAT1B1 and OAT1B3 (IC50s = 1.5 and 3.27 µM, respectively)
Catalog No. 50-299-4938