Learn More
TARGETMOL CHEMICALS INC DACOMITINIB 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T248310MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dacomitinib (PF299)(PF299804; PF-00299804) is a highly selective orally bioavailable small-molecule inhibitor of the HER family of tyrosine kinases with IC50 of 6 45.7 and 73.7 nM for EGFR ERBB2 and ERBB4 respectively. purity: 99%
Catalog No. 50-257-7110