Learn More
Abcam DAF-2 DA, Fluorescent nitric o x ide probe, 5MG
Supplier:  Abcam AB1452835MG
Encompass_Preferred
MW 446.41 g/mol, Purity >98%. Cell-permeable fluorescent nitric oxide (NO) indicator. Deacetylated by intracellular esterases to DAF-2 (ab145269). DAF-2 reacts with NO to yield the highly fluorescent triazolofluorescein (DAF-2T) (ab145473).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3261