Learn More
AdipoGen DAF-2T Solution 5 mM in DMSO
SDP
Supplier:  AdipoGen CDXD1234M001
Encompass_Preferred
Chemical. CAS 261351-48-8. Formula C28H29N5O3. MW 483.6. Reference compound for the fluorescent nitric oxide NO probe DAR-2.
Catalog No. 50-256-1006